missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha'-Dibromo-o-xylene 98.0+%, TCI America™

Catalog No. p-7126088
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D021425G 25 g
D0214100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D021425G Supplier TCI America Supplier No. D021425G
Only null left

Chemical Identifiers

CAS 91-13-4
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
MDL Number MFCD00000175
InChI Key KGKAYWMGPDWLQZ-UHFFFAOYSA-N
Synonym 1,2-bis bromomethyl benzene, o-xylylene dibromide, alpha,alpha'-dibromo-o-xylene, benzene, 1,2-bis bromomethyl, o-bis bromomethyl benzene, o-xylylene bromide, 1,2-dibenzyl bromide, alpha,alpha'-dibromo-o-xylol, a,a'-dibromo-o-xylene, ccris 1776
PubChem CID 66665
IUPAC Name 1,2-bis(bromomethyl)benzene
SMILES C1=CC=C(C(=C1)CBr)CBr

Specifications

Melting Point 93°C
Color White
Boiling Point 140°C
Quantity 25 g
UN Number 2928
Formula Weight 263.96
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha′-Dibromo-o-xylene