missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha,alpha',alpha',4,5-Hexabromo-o-xylene 98.0+%, TCI America™

Catalog No. p-7130749
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08311G 1 g
H08315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08311G Supplier TCI America Supplier No. H08311G
Only null left

Chemical Identifiers

CAS 13209-20-6
Molecular Formula C8H4Br6
Molecular Weight (g/mol) 579.544
MDL Number MFCD00191445
InChI Key QPLVNMIEWOGDBY-UHFFFAOYSA-N
PubChem CID 12186451
IUPAC Name 1,2-dibromo-4,5-bis(dibromomethyl)benzene
SMILES C1=C(C(=CC(=C1Br)Br)C(Br)Br)C(Br)Br

Specifications

Melting Point 135°C
Color White-Yellow
Quantity 1 g
UN Number 1759
Formula Weight 579.54
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha,alpha′,alpha′,4,5-Hexabromo-o-xylene