accessibility menu, dialog, popup

UserName

BrBrBrBrBrBr

CAS RN 13209-20-6

IUPAC Name: 1,2-dibromo-4,5-bis(dibromomethyl)benzene
Synonyms: 1,2-dibromo-4,5-bis(dibromomethyl)benzene
Molecular Weight (g/mol): 579.54
Molecular Formula: C8H4Br6
InChi Key: QPLVNMIEWOGDBY-UHFFFAOYSA-N
SMILES: BrC(Br)C1=CC(Br)=C(Br)C=C1C(Br)Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results