missing translation for 'onlineSavingsMsg'
Learn More

alpha-2-Chloroisodurene, 98%

Catalog No. AC159780250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159780250 25 g Glass bottle
AC159780050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC159780250 Supplier Thermo Scientific Chemicals Supplier No. 159780250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1585-16-6 | C10H13Cl | 168.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1585-16-6
Molecular Formula C10H13Cl
Molecular Weight (g/mol) 168.66
MDL Number MFCD00000901
InChI Key UNRGEIXQCZHICP-UHFFFAOYSA-N
Synonym 2,4,6-trimethylbenzyl chloride, 2-chloromethyl-1,3,5-trimethylbenzene, alpha-2-chloroisodurene, benzene, 2-chloromethyl-1,3,5-trimethyl, 2,4,6-trimethyl benzyl chloride, alpha2-chloroisodurene, chloromethyl mesitylene, 2-chloromethyl-1,3,5-trimethyl-benzene, az-chloroisodurene, a-2-chloroisodurene
PubChem CID 74108
IUPAC Name 2-(chloromethyl)-1,3,5-trimethylbenzene
SMILES CC1=CC(C)=C(CCl)C(C)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 34°C to 38°C
Color White
Boiling Point 114°C to 115°C (10.0 mmHg)
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3C6H2CH2Cl
Quantity 25 g
Solubility Information Solubility in water: insoluble
Formula Weight 168.67
Percent Purity 98%
Physical Form Fused Solid, Crystals, or Chunks
Chemical Name or Material α-2-Chloroisodurene, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep only in
EINECSNumber 216-440-3
TSCA TSCA

RUO – Research Use Only