Learn More
Allyltriphenylphosphonium chloride, 99%
CAS: 18480-23-4 | C21H20ClP | 338.81 g/mol
$101.51 - $1361.05
Chemical Identifiers
| CAS | 18480-23-4 |
|---|---|
| Molecular Formula | C21H20ClP |
| Molecular Weight (g/mol) | 338.81 |
| MDL Number | MFCD00031542 |
| InChI Key | FKMJROWWQOJRJX-UHFFFAOYSA-M |
| Synonym | allyltriphenylphosphonium chloride, allyl triphenylphosphonium chloride, triphenyl prop-2-enyl phosphanium chloride, triphenyl prop-2-en-1-yl phosphanium chloride, acmc-1buz2, allyltriphenylphosphoniumchloride, triphenyl prop-2-enyl phosphonium chloride, phosphonium,triphenyl-2-propen-1-yl-, chloride 1:1 |
| PubChem CID | 2734120 |
| SMILES | [Cl-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1601214
|
Thermo Scientific Chemicals
A1601214 |
25 g |
Each for $101.51
|
|
|||||
|
AAA1601222
|
Thermo Scientific Chemicals
A1601222 |
100 g |
Each for $347.77
|
|
|||||
|
AAA1601236
|
Thermo Scientific Chemicals
A1601236 |
500 g |
Each for $1,361.05
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 18480-23-4 | |
| 338.81 | |
| FKMJROWWQOJRJX-UHFFFAOYSA-M | |
| 2734120 |
| C21H20ClP | |
| MFCD00031542 | |
| allyltriphenylphosphonium chloride, allyl triphenylphosphonium chloride, triphenyl prop-2-enyl phosphanium chloride, triphenyl prop-2-en-1-yl phosphanium chloride, acmc-1buz2, allyltriphenylphosphoniumchloride, triphenyl prop-2-enyl phosphonium chloride, phosphonium,triphenyl-2-propen-1-yl-, chloride 1:1 | |
| [Cl-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
| 18480-23-4 | |
| C21H20ClP | |
| MFCD00031542 | |
| Hygroscopic | |
| FKMJROWWQOJRJX-UHFFFAOYSA-M | |
| triphenyl(prop-2-enyl)phosphanium;chloride | |
| 2734120 | |
| 99% |
| 230°C to 232°C | |
| (CH2=CHCH2)(C6H5)3PCl | |
| 25 g | |
| allyltriphenylphosphonium chloride, allyl triphenylphosphonium chloride, triphenyl prop-2-enyl phosphanium chloride, triphenyl prop-2-en-1-yl phosphanium chloride, acmc-1buz2, allyltriphenylphosphoniumchloride, triphenyl prop-2-enyl phosphonium chloride, phosphonium,triphenyl-2-propen-1-yl-, chloride 1:1 | |
| [Cl-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 338.81 | |
| 338.82 | |
| Allyltriphenylphosphonium chloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 242-368-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only