CAS RN 18480-23-4
IUPAC Name:
triphenyl(prop-2-en-1-yl)phosphanium chloride
Synonyms:
triphenyl(prop-2-en-1-yl)phosphanium chloride
Molecular Weight (g/mol):
338.81
Molecular Formula:
C21H20ClP
InChi Key:
FKMJROWWQOJRJX-UHFFFAOYSA-M
SMILES:
[Cl-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Allyltriphenylphosphonium chloride, 99%
CAS: 18480-23-4 Molecular Formula: C21H20ClP Molecular Weight (g/mol): 338.81 MDL Number: MFCD00031542 InChI Key: FKMJROWWQOJRJX-UHFFFAOYSA-M Synonym: allyltriphenylphosphonium chloride,allyl triphenylphosphonium chloride,triphenyl prop-2-enyl phosphanium chloride,triphenyl prop-2-en-1-yl phosphanium chloride,acmc-1buz2,allyltriphenylphosphoniumchloride,triphenyl prop-2-enyl phosphonium chloride,phosphonium,triphenyl-2-propen-1-yl-, chloride 1:1 PubChem CID: 2734120 SMILES: [Cl-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1