missing translation for 'onlineSavingsMsg'
Learn More

Allyltriphenylphosphonium chloride, 99%

Catalog No. AAA1601214
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1601214 25 g
AAA1601222 100 g
AAA1601236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1601214 Supplier Thermo Scientific Chemicals Supplier No. A1601214

CAS: 18480-23-4 | C21H20ClP | 338.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18480-23-4
Molecular Formula C21H20ClP
Molecular Weight (g/mol) 338.81
MDL Number MFCD00031542
InChI Key FKMJROWWQOJRJX-UHFFFAOYSA-M
Synonym allyltriphenylphosphonium chloride, allyl triphenylphosphonium chloride, triphenyl prop-2-enyl phosphanium chloride, triphenyl prop-2-en-1-yl phosphanium chloride, acmc-1buz2, allyltriphenylphosphoniumchloride, triphenyl prop-2-enyl phosphonium chloride, phosphonium,triphenyl-2-propen-1-yl-, chloride 1:1
PubChem CID 2734120
SMILES [Cl-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 230°C to 232°C
Linear Formula (CH2=CHCH2)(C6H5)3PCl
Quantity 25 g
Sensitivity Hygroscopic
IUPAC Name triphenyl(prop-2-enyl)phosphanium;chloride
Formula Weight 338.82
Percent Purity 99%
Chemical Name or Material Allyltriphenylphosphonium chloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 242-368-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only