missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyltrimethylsilane 98.0+%, TCI America™
$111.84 - $643.10
Chemical Identifiers
| CAS | 762-72-1 |
|---|---|
| Molecular Formula | C6H14Si |
| Molecular Weight (g/mol) | 114.263 |
| MDL Number | MFCD00008635 |
| InChI Key | HYWCXWRMUZYRPH-UHFFFAOYSA-N |
| Synonym | allyltrimethylsilane, silane, trimethyl-2-propenyl, trimethylallylsilane, silane, allyltrimethyl, allyl trimethylsilane, unii-8b84c337vf, ccris 2649, allyl trimethyl silane, 3-trimethylsilyl propene, allytrimethylsilane |
| PubChem CID | 69808 |
| IUPAC Name | trimethyl(prop-2-enyl)silane |
| SMILES | C[Si](C)(C)CC=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A072925ML
|
TCI America
A072925ML |
25 mL |
Each for $111.84
|
|
|||||
|
A0729250ML
|
TCI America
A0729250ML |
250 mL |
Each for $643.10
|
|
|||||
Chemical Identifiers
| 762-72-1 | |
| 114.263 | |
| HYWCXWRMUZYRPH-UHFFFAOYSA-N | |
| 69808 | |
| C[Si](C)(C)CC=C |
| C6H14Si | |
| MFCD00008635 | |
| allyltrimethylsilane, silane, trimethyl-2-propenyl, trimethylallylsilane, silane, allyltrimethyl, allyl trimethylsilane, unii-8b84c337vf, ccris 2649, allyl trimethyl silane, 3-trimethylsilyl propene, allytrimethylsilane | |
| trimethyl(prop-2-enyl)silane |
Specifications
| 762-72-1 | |
| 83°C | |
| MFCD00008635 | |
| 1993 | |
| HYWCXWRMUZYRPH-UHFFFAOYSA-N | |
| trimethyl(prop-2-enyl)silane | |
| 69808 | |
| ≥98.0% (GC) | |
| Allyltrimethylsilane |
| Colorless | |
| C6H14Si | |
| 25 mL | |
| allyltrimethylsilane, silane, trimethyl-2-propenyl, trimethylallylsilane, silane, allyltrimethyl, allyl trimethylsilane, unii-8b84c337vf, ccris 2649, allyl trimethyl silane, 3-trimethylsilyl propene, allytrimethylsilane | |
| C[Si](C)(C)CC=C | |
| 114.263 | |
| 114.26 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-3737
TSCA : Yes