missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyltrimethylsilane 98.0+%, TCI America™
Supplier: TCI America A0729250ML
Specifications
| Allyltrimethylsilane | |
| Colorless | |
| C6H14Si | |
| 250 mL | |
| allyltrimethylsilane, silane, trimethyl-2-propenyl, trimethylallylsilane, silane, allyltrimethyl, allyl trimethylsilane, unii-8b84c337vf, ccris 2649, allyl trimethyl silane, 3-trimethylsilyl propene, allytrimethylsilane | |
| C[Si](C)(C)CC=C | |
| 114.263 | |
| 114.26 | |
| Liquid |
| 762-72-1 | |
| 83°C | |
| MFCD00008635 | |
| 1993 | |
| HYWCXWRMUZYRPH-UHFFFAOYSA-N | |
| trimethyl(prop-2-enyl)silane | |
| 69808 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 762-72-1 | |
| 114.263 | |
| HYWCXWRMUZYRPH-UHFFFAOYSA-N | |
| 69808 | |
| C[Si](C)(C)CC=C |
| C6H14Si | |
| MFCD00008635 | |
| allyltrimethylsilane, silane, trimethyl-2-propenyl, trimethylallylsilane, silane, allyltrimethyl, allyl trimethylsilane, unii-8b84c337vf, ccris 2649, allyl trimethyl silane, 3-trimethylsilyl propene, allytrimethylsilane | |
| trimethyl(prop-2-enyl)silane |
Safety and Handling
EINECSNumber : (2)-3737
TSCA : Yes