missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allylpentafluorobenzene 98.0+%, TCI America™
$74.56 - $253.51
Chemical Identifiers
| CAS | 1736-60-3 |
|---|---|
| Molecular Formula | C9H5F5 |
| Molecular Weight (g/mol) | 208.13 |
| MDL Number | MFCD00000301 |
| InChI Key | YBDBTBVNQQBHGJ-UHFFFAOYSA-N |
| Synonym | allylpentafluorobenzene, benzene, allylpentafluoro, 3-pentafluorophenyl-1-propene, 2,3,4,5,6-pentafluoro allylbenzene, benzene, pentafluoro-2-propenyl, 1,2,3,4,5-pentafluoro-6-prop-2-en-1-yl benzene, allyl pentafluorobenzene, pubchem13366, acmc-1bth5 |
| PubChem CID | 74435 |
| IUPAC Name | 1,2,3,4,5-pentafluoro-6-(prop-2-en-1-yl)benzene |
| SMILES | FC1=C(F)C(F)=C(CC=C)C(F)=C1F |
Chemical Identifiers
| 1736-60-3 | |
| 208.13 | |
| YBDBTBVNQQBHGJ-UHFFFAOYSA-N | |
| 74435 | |
| FC1=C(F)C(F)=C(CC=C)C(F)=C1F |
| C9H5F5 | |
| MFCD00000301 | |
| allylpentafluorobenzene, benzene, allylpentafluoro, 3-pentafluorophenyl-1-propene, 2,3,4,5,6-pentafluoro allylbenzene, benzene, pentafluoro-2-propenyl, 1,2,3,4,5-pentafluoro-6-prop-2-en-1-yl benzene, allyl pentafluorobenzene, pubchem13366, acmc-1bth5 | |
| 1,2,3,4,5-pentafluoro-6-(prop-2-en-1-yl)benzene |
Specifications
| 1736-60-3 | |
| 149°C | |
| MFCD00000301 | |
| 1993 | |
| YBDBTBVNQQBHGJ-UHFFFAOYSA-N | |
| 1,2,3,4,5-pentafluoro-6-(prop-2-en-1-yl)benzene | |
| 74435 | |
| ≥98.0% (GC) | |
| Allylpentafluorobenzene |
| Yellow | |
| C9H5F5 | |
| 5 g | |
| allylpentafluorobenzene, benzene, allylpentafluoro, 3-pentafluorophenyl-1-propene, 2,3,4,5,6-pentafluoro allylbenzene, benzene, pentafluoro-2-propenyl, 1,2,3,4,5-pentafluoro-6-prop-2-en-1-yl benzene, allyl pentafluorobenzene, pubchem13366, acmc-1bth5 | |
| FC1=C(F)C(F)=C(CC=C)C(F)=C1F | |
| 208.13 | |
| 208.13 | |
| Liquid |
Safety and Handling
TSCA : No