accessibility menu, dialog, popup

UserName

FFFCH2FF

CAS RN 1736-60-3

IUPAC Name: 1,2,3,4,5-pentafluoro-6-(prop-2-en-1-yl)benzene
Synonyms: 1,2,3,4,5-pentafluoro-6-(prop-2-en-1-yl)benzene
Molecular Weight (g/mol): 208.13
Molecular Formula: C9H5F5
InChi Key: YBDBTBVNQQBHGJ-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(CC=C)C(F)=C1F
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)