missing translation for 'onlineSavingsMsg'
Learn More

Allylpentafluorobenzene 98.0+%, TCI America™

Catalog No. A16115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16115G 5 g
A161125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16115G Supplier TCI America Supplier No. A16115G
Only null left

Chemical Identifiers

CAS 1736-60-3
Molecular Formula C9H5F5
Molecular Weight (g/mol) 208.13
MDL Number MFCD00000301
InChI Key YBDBTBVNQQBHGJ-UHFFFAOYSA-N
Synonym allylpentafluorobenzene, benzene, allylpentafluoro, 3-pentafluorophenyl-1-propene, 2,3,4,5,6-pentafluoro allylbenzene, benzene, pentafluoro-2-propenyl, 1,2,3,4,5-pentafluoro-6-prop-2-en-1-yl benzene, allyl pentafluorobenzene, pubchem13366, acmc-1bth5
PubChem CID 74435
IUPAC Name 1,2,3,4,5-pentafluoro-6-(prop-2-en-1-yl)benzene
SMILES FC1=C(F)C(F)=C(CC=C)C(F)=C1F

Specifications

Color Yellow
Boiling Point 149°C
Quantity 5 g
UN Number 1993
Formula Weight 208.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allylpentafluorobenzene