missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyl Propyl Sulfide 97.0+%, TCI America™
Supplier: TCI America A106125ML
Specifications
| Allyl Propyl Sulfide | |
| Yellow | |
| C6H12S | |
| 25 mL | |
| JMLIYQJQKZYHDQ-UHFFFAOYSA-N | |
| 1-prop-2-enylsulfanylpropane | |
| 98781 | |
| ≥97.0% (GC) |
| 27817-67-0 | |
| 140°C | |
| MFCD00015220 | |
| 1993 | |
| CCCSCC=C | |
| 116.222 | |
| 116.22 | |
| Liquid |
Chemical Identifiers
| 27817-67-0 | |
| 116.222 | |
| JMLIYQJQKZYHDQ-UHFFFAOYSA-N | |
| 1-prop-2-enylsulfanylpropane |
| C6H12S | |
| MFCD00015220 | |
| 98781 | |
| CCCSCC=C |
Safety and Handling
TSCA : No