accessibility menu, dialog, popup

UserName

CAS RN 27817-67-0

IUPAC Name: 3-(propylsulfanyl)prop-1-ene
Synonyms: 3-(propylsulfanyl)prop-1-ene
Molecular Weight (g/mol): 116.22
Molecular Formula: C6H12S
InChi Key: JMLIYQJQKZYHDQ-UHFFFAOYSA-N
SMILES: CCCSCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results