missing translation for 'onlineSavingsMsg'
Learn More

Allyl Propyl Disulfide 88.0+%, TCI America™

Catalog No. a19405g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A19405G Supplier TCI America Supplier No. A19405G
Only null left

Chemical Identifiers

CAS 2179-59-1
Molecular Formula C6H12S2
Molecular Weight (g/mol) 148.282
MDL Number MFCD00040072
InChI Key FCSSPCOFDUKHPV-UHFFFAOYSA-N
PubChem CID 16591
IUPAC Name 1-(prop-2-enyldisulfanyl)propane
SMILES CCCSSCC=C

Specifications

Color Yellow
Boiling Point 69°C
Quantity 5 g
UN Number 1993
Formula Weight 148.28
Percent Purity ≥88.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Propyl Disulfide
RTECSNumber JO0350000
TSCA No
Recommended Storage Refrigerator