accessibility menu, dialog, popup

UserName

CAS RN 2179-59-1

IUPAC Name: 3-(propyldisulfanyl)prop-1-ene
Synonyms: allyl propyl disulfide disulfide, 2-propenyl propyl
Molecular Weight (g/mol): 148.28
Molecular Formula: C6H12S2
InChi Key: FCSSPCOFDUKHPV-UHFFFAOYSA-N
SMILES: CCCSSCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results