missing translation for 'onlineSavingsMsg'
Learn More

Allyl Phenyl Ether 95.0+%, TCI America™

Catalog No. p-7116779
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A023425G 25 g
A0234100G 100 g
A0234500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A023425G Supplier TCI America Supplier No. A023425G
Only null left

Chemical Identifiers

CAS 1746-13-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00008644
InChI Key POSICDHOUBKJKP-UHFFFAOYSA-N
Synonym allyl phenyl ether, allyloxy benzene, allyloxybenzene, benzene, 2-propenyloxy, allyl phenoxylate, phenyl allyl ether, ether, allyl phenyl, 3-phenoxypropene, phenylpropenyl ether, phenyl 2-propenyl ether
PubChem CID 74458
IUPAC Name prop-2-enoxybenzene
SMILES C=CCOC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 195°C
Quantity 25 g
Formula Weight 134.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Phenyl Ether
EINECSNumber (3)-2837
RTECSNumber DA8575000
TSCA Yes