accessibility menu, dialog, popup

UserName

CAS RN 1746-13-0

IUPAC Name: (prop-2-en-1-yloxy)benzene
Synonyms: allyl phenyl ether
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: POSICDHOUBKJKP-UHFFFAOYSA-N
SMILES: C=CCOC1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)