missing translation for 'onlineSavingsMsg'
Learn More

Allyl phenyl ether, 99%

Catalog No. AAL0335922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0335922 100 g
AAL0335914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0335922 Supplier Thermo Scientific Chemicals Supplier No. L0335922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1746-13-0 | C9H10O | 134.178 g/mol

Allyl phenyl ether is an pharmaceutical and OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl phenyl ether is an pharmaceutical and OLED intermediate.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents, acids, bases. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1746-13-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00008644
InChI Key POSICDHOUBKJKP-UHFFFAOYSA-N
Synonym allyl phenyl ether, allyloxy benzene, allyloxybenzene, benzene, 2-propenyloxy, allyl phenoxylate, phenyl allyl ether, ether, allyl phenyl, 3-phenoxypropene, phenylpropenyl ether, phenyl 2-propenyl ether
PubChem CID 74458
IUPAC Name prop-2-enoxybenzene
SMILES C=CCOC1=CC=CC=C1

Specifications

Density 0.979
Boiling Point 191°C to 192°C
Flash Point 62°C (143°F)
Odor Characteristic
Refractive Index 1.521
Quantity 100 g
Beilstein 1905622
Solubility Information Insoluble in water.
Formula Weight 134.18
Percent Purity 99%
Chemical Name or Material Allyl phenyl ether
Show More Show Less
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 217-125-3
RTECSNumber DA8575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only