missing translation for 'onlineSavingsMsg'
Learn More

Allyl Phenyl Carbonate 97.0+%, TCI America™

Catalog No. A23035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23035G 5 g
A230325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23035G Supplier TCI America Supplier No. A23035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16308-68-2
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD07784342
InChI Key ORUWSEKEVGOAQR-UHFFFAOYSA-N
Synonym Carbonic Acid Allyl Phenyl Ester
PubChem CID 11412694
IUPAC Name phenyl prop-2-en-1-yl carbonate
SMILES C=CCOC(=O)OC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 78°C
Quantity 5 g
Formula Weight 178.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Phenyl Carbonate