accessibility menu, dialog, popup

UserName

H2COOO

CAS RN 16308-68-2

IUPAC Name: phenyl prop-2-en-1-yl carbonate
Synonyms: phenyl prop-2-en-1-yl carbonate
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: ORUWSEKEVGOAQR-UHFFFAOYSA-N
SMILES: C=CCOC(=O)OC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results