missing translation for 'onlineSavingsMsg'
Learn More

Allyl Methyl Disulfide 90.0+%, TCI America™

Catalog No. a19395g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A19395G Supplier TCI America Supplier No. A19395G
Only null left

Chemical Identifiers

CAS 2179-58-0
Molecular Formula C4H8S2
Molecular Weight (g/mol) 120.228
MDL Number MFCD00040023
InChI Key XNZOTQPMYMCTBZ-UHFFFAOYSA-N
PubChem CID 62434
ChEBI CHEBI:6854
IUPAC Name 3-(methyldisulfanyl)prop-1-ene
SMILES CSSCC=C

Specifications

Color Yellow
Quantity 5 g
UN Number 1993
Formula Weight 120.23
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Methyl Disulfide
TSCA No
Recommended Storage Refrigerator