accessibility menu, dialog, popup

UserName

CAS RN 2179-58-0

IUPAC Name: 3-(methyldisulfanyl)prop-1-ene
Synonyms: allyl methyl disulfide methyl 2-propenyl disulfide methylallyl disulphide 3-(methyldithio)prop-1-ene allyl methyl disulphide disulfide, allyl methyl disulfide, methyl 2-propenyl methyl allyl disulfide
Molecular Weight (g/mol): 120.23
Molecular Formula: C4H8S2
InChi Key: XNZOTQPMYMCTBZ-UHFFFAOYSA-N
SMILES: CSSCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results