missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyl Iodide (stabilized with Copper chip) 98.0+%, TCI America™
Supplier: TCI America I007025G
Specifications
| Allyl Iodide (stabilized with Copper chip) | |
| -99°C | |
| 102°C | |
| MFCD00001094 | |
| 1723 | |
| HFEHLDPGIKPNKL-UHFFFAOYSA-N | |
| 3-iodoprop-1-ene | |
| 11166 | |
| ≥98.0% (GC) |
| 556-56-9 | |
| Brown-Yellow | |
| C3H5I | |
| 25 g | |
| allyl iodide, 1-propene, 3-iodo, 3-iodopropene, 3-iodo-1-propene, 3-iodopropylene, propene, 3-iodo, allyliodide, unii-46830qoa4d, 3-iodo-propene, 3-iodanylprop-1-ene | |
| ICC=C | |
| 167.98 | |
| 167.98 | |
| Liquid |
Chemical Identifiers
| 556-56-9 | |
| 167.98 | |
| HFEHLDPGIKPNKL-UHFFFAOYSA-N | |
| 11166 | |
| ICC=C |
| C3H5I | |
| MFCD00001094 | |
| allyl iodide, 1-propene, 3-iodo, 3-iodopropene, 3-iodo-1-propene, 3-iodopropylene, propene, 3-iodo, allyliodide, unii-46830qoa4d, 3-iodo-propene, 3-iodanylprop-1-ene | |
| 3-iodoprop-1-ene |
Safety and Handling
RTECSNumber : UD0450000
TSCA : Yes
Recommended Storage : Refrigerator