accessibility menu, dialog, popup

UserName

IH2C

CAS RN 556-56-9

IUPAC Name: 3-iodoprop-1-ene
Synonyms: allyl iodide
Molecular Weight (g/mol): 167.98
Molecular Formula: C3H5I
InChi Key: HFEHLDPGIKPNKL-UHFFFAOYSA-N
SMILES: ICC=C
Boiling Point 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)