missing translation for 'onlineSavingsMsg'
Learn More

Allyl Iodide (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. p-7131194
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I00705G 5 g
I007025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I00705G Supplier TCI America Supplier No. I00705G
Only null left

Chemical Identifiers

CAS 556-56-9
Molecular Formula C3H5I
Molecular Weight (g/mol) 167.98
MDL Number MFCD00001094
InChI Key HFEHLDPGIKPNKL-UHFFFAOYSA-N
Synonym allyl iodide, 1-propene, 3-iodo, 3-iodopropene, 3-iodo-1-propene, 3-iodopropylene, propene, 3-iodo, allyliodide, unii-46830qoa4d, 3-iodo-propene, 3-iodanylprop-1-ene
PubChem CID 11166
IUPAC Name 3-iodoprop-1-ene
SMILES ICC=C

Specifications

Melting Point -99°C
Color Brown-Yellow
Boiling Point 102°C
Quantity 5 g
UN Number 1723
Formula Weight 167.98
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Iodide (stabilized with Copper chip)
RTECSNumber UD0450000
TSCA Yes
Recommended Storage Refrigerator