missing translation for 'onlineSavingsMsg'
Learn More

Allyl chloride, 98%, stab. with propylene oxide, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A14330AP

 View more versions of this product

Catalog No. AAA14330AP


Only null left
Explore available promotions

Description

Description

Allyl chloride is used as an alkylating agent. It is used in treatment with sodium bis(trimethylsilyl)amide gives the strained, highly reactive alkene cyclopropene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl chloride is used as an alkylating agent. It is used in treatment with sodium bis(trimethylsilyl)amide gives the strained, highly reactive alkene cyclopropene.

Solubility
Soluble in water (3600 mg/L at 20°C), ethanol, ether, petroleum, acetone, chloroform, benzene, methanol and 2-prop.

Notes
Store at -20°C. Keep tightly closed in a dry and cool place. Keep in properly labeled containers. Keep containers tightly closed in a cool, well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents, boron trifluoride, sulfuric acid and nitric acid
Specifications

Specifications

Allyl chloride
107-05-1
0.939
−28°C (−18°F)
C3H5Cl
CH2=CHCH2Cl
500 mL
635704
allyl chloride, 3-chloro-1-propene, 1-propene, 3-chloro, 3-chloropropylene, 3-chloropropene, chlorallylene, allylchlorid, 2-propenyl chloride, 1-chloro-2-propene, allylchloride
OSDWBNJEKMUWAV-UHFFFAOYSA-N
3-chloroprop-1-ene
7850
76.53
stabilized with propylene oxide
-135°C
44°C to 46°C
Sweet
1.4155
MFCD00000984
UN1100
14,289
Soluble in water (3600mg/L at 20°C),ethanol,ether,petroleum,acetone,chloroform,benzene,methanol and 2-prop.
C=CCCl
76.523
CHEBI:82379
98%

Chemical Identifiers

107-05-1
MFCD00000984
allyl chloride, 3-chloro-1-propene, 1-propene, 3-chloro, 3-chloropropylene, 3-chloropropene, chlorallylene, allylchlorid, 2-propenyl chloride, 1-chloro-2-propene, allylchloride
CHEBI:82379
C3H5Cl
OSDWBNJEKMUWAV-UHFFFAOYSA-N
7850
3-chloroprop-1-ene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H341-H351-H373-H302-H312-H332-H315-H319-H335
Highly flammable liquid and vapour.
Suspected of causing genetic defects.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P363-P370+P378q-P5

H225-H302+H312+H332-H315-H319-H335-H341-H351-H373

DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: ALLYL CHLORIDE

EINECSNumber : 203-457-6

RTECSNumber : UC7350000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only