missing translation for 'onlineSavingsMsg'
Learn More

Allyl chloride, 98%, stab. with propylene oxide, Thermo Scientific Chemicals

Catalog No. p-7021833
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA14330AE 100 mL
AAA14330AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA14330AE Supplier Thermo Scientific Chemicals Supplier No. A14330AE
Only null left

Allyl chloride is used as an alkylating agent. It is used in treatment with sodium bis(trimethylsilyl)amide gives the strained, highly reactive alkene cyclopropene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl chloride is used as an alkylating agent. It is used in treatment with sodium bis(trimethylsilyl)amide gives the strained, highly reactive alkene cyclopropene.

Solubility
Soluble in water (3600 mg/L at 20°C), ethanol, ether, petroleum, acetone, chloroform, benzene, methanol and 2-prop.

Notes
Store at -20°C. Keep tightly closed in a dry and cool place. Keep in properly labeled containers. Keep containers tightly closed in a cool, well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents, boron trifluoride, sulfuric acid and nitric acid

Chemical Identifiers

CAS 107-05-1
Molecular Formula C3H5Cl
MDL Number MFCD00000984
InChI Key OSDWBNJEKMUWAV-UHFFFAOYSA-N
Synonym allyl chloride, 3-chloro-1-propene, 1-propene, 3-chloro, 3-chloropropylene, 3-chloropropene, chlorallylene, allylchlorid, 2-propenyl chloride, 1-chloro-2-propene, allylchloride
PubChem CID 7850
ChEBI CHEBI:82379
IUPAC Name 3-chloroprop-1-ene

Specifications

Melting Point -135°C
Density 0.939
Boiling Point 44°C to 46°C
Flash Point −28°C (−18°F)
Odor Sweet
Refractive Index 1.4155
Linear Formula CH2=CHCH2Cl
Quantity 100 mL
UN Number UN1100
Beilstein 635704
Merck Index 14,289
Solubility Information Soluble in water (3600mg/L at 20°C),ethanol,ether,petroleum,acetone,chloroform,benzene,methanol and 2-prop.
SMILES C=CCCl
Molecular Weight (g/mol) 76.523
Formula Weight 76.53
Percent Purity 98%
Chemical Name or Material Allyl chloride, stabilized with propylene oxide
Show More Show Less
Hazard Category H225-H302+H312+H332-H315-H319-H335-H341-H351-H373
Hazard Statement GHS H Statement
H225-H341-H351-H373-H302-H312-H332-H315-H319-H335
Highly flammable liquid and vapour.
Suspected of causing genetic defects.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P363-P370+P378q-P5
DOTInformation Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: ALLYL CHLORIDE
EINECSNumber 203-457-6
RTECSNumber UC7350000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only