missing translation for 'onlineSavingsMsg'
Learn More

Allyl Bromide 98.0+%, TCI America™

Catalog No. B064325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B064325G 25 g
B0643500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B064325G Supplier TCI America Supplier No. B064325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106-95-6
Molecular Formula C3H5Br
Molecular Weight (g/mol) 120.977
MDL Number MFCD00000244
InChI Key BHELZAPQIKSEDF-UHFFFAOYSA-N
Synonym allyl bromide, 3-bromopropene, 1-propene, 3-bromo, 3-bromopropylene, 3-bromo-1-propene, bromallylene, 2-propenyl bromide, 1-bromo-2-propene, propene, 3-bromo, allylbromide
PubChem CID 7841
IUPAC Name 3-bromoprop-1-ene
SMILES C=CCBr

Specifications

Color Yellow
Boiling Point 71°C
Quantity 25 g
UN Number 1099
Formula Weight 120.98
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Bromide
EINECSNumber (2)-0107&(9)-0129
RTECSNumber UC7090000
TSCA Yes
Recommended Storage Refrigerator