missing translation for 'onlineSavingsMsg'
Learn More

Allyl bromide, 99%, stabilized

Catalog No. AC102900100
Change view
Click to view available options
Quantity:
250 mL
500 mL
5 L
10 L
Packaging:
Glass bottle
Metal Can
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102900100 10 L Plastic drum
AC102902500 250 mL Glass bottle
AC102905000 500 mL Glass bottle
AC102900050 5 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC102900100 Supplier Thermo Scientific Chemicals Supplier No. 102900100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 106-95-6 | C3H5Br | 120.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent

Chemical Identifiers

CAS 106-95-6 , 75-56-9
Molecular Formula C3H5Br
Molecular Weight (g/mol) 120.98
MDL Number MFCD00000244
InChI Key BHELZAPQIKSEDF-UHFFFAOYSA-N
Synonym allyl bromide, 3-bromopropene, 1-propene, 3-bromo, 3-bromopropylene, 3-bromo-1-propene, bromallylene, 2-propenyl bromide, 1-bromo-2-propene, propene, 3-bromo, allylbromide
PubChem CID 7841
IUPAC Name 3-bromoprop-1-ene
SMILES C=CCBr

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -119.0°C
Color Colorless
Density 1.3900g/mL
Boiling Point 70.0°C to 71.0°C
Flash Point −1°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Odor The liquid has a pungent odor
Packaging Plastic drum
Refractive Index 1.4680 to 1.4700
Linear Formula H2C=CHCH2Br
Quantity 10 L
Beilstein 01,201
Fieser 13,219
Specific Gravity 1.39
Merck Index 15,279
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol, chloroform, ether,, carbontetrachloride and carbon disulfide
Stabilizer 300ppm propylene oxide
Formula Weight 120.98
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Allyl bromide, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
Toxic if swallowed.
Very toxic to aquatic life.
Highly flammable liquid and vapor.
May cause genetic defects.
May cause cancer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
EINECSNumber 203-446-6

RUO – Research Use Only