missing translation for 'onlineSavingsMsg'
Learn More

Allyl bromide, 99%, stab. with 300-1000ppm Propylene oxide

CAS: 106-95-6 | C3H5Br | 120.977 g/mol

Supplier:  Thermo Scientific Chemicals A117660B

 View more versions of this product

Catalog No. AAA117660B

Only null left
Explore available promotions

Description

Description

Allyl bromide is used as an alkylating agent in the synthesis of pharmaceuticals, polymers, adhesives, perfumes, biochemicals and other allylic compounds. It is used as precursor for the preparation of allyliczinc bromide by reacting it with zinc. It is also used in the preparation of allylethers like allyl decyl ether, allyl benzyl ether and allyl geranyl ether. It is also used in the preparation of R enantiomer of allyl phenyl carbinol (APC) such as 1-phenyl-3-butene, which is a valuable intermediate for drugs and agro-chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl bromide is used as an alkylating agent in the synthesis of pharmaceuticals, polymers, adhesives, perfumes, biochemicals and other allylic compounds. It is used as precursor for the preparation of allyliczinc bromide by reacting it with zinc. It is also used in the preparation of allylethers like allyl decyl ether, allyl benzyl ether and allyl geranyl ether. It is also used in the preparation of R enantiomer of allyl phenyl carbinol (APC) such as 1-phenyl-3-butene, which is a valuable intermediate for drugs and agro-chemicals.

Solubility
Slightly miscible with water.

Notes
Moisture and light sensitive. Incompatible with oxidizing agents, alkali metals, alkaline earth metals, amides, amines and powdered metals.
Specifications

Specifications

Allyl bromide
106-95-6
1.398
−2°C (28°F)
Pungent
1.47
MFCD00000244
UN1099
14,288
Slightly miscible with water.
C=CCBr
120.977
120.98
Liquid
Stabilized with 300 to 1000ppm Propylene oxide
-119°C
70°C to 71°C
99%
C3H5Br
CH2=CHCH2Br
1000 g
Light sensitive
allyl bromide, 3-bromopropene, 1-propene, 3-bromo, 3-bromopropylene, 3-bromo-1-propene, bromallylene, 2-propenyl bromide, 1-bromo-2-propene, propene, 3-bromo, allylbromide
BHELZAPQIKSEDF-UHFFFAOYSA-N
3-bromoprop-1-ene
7841
99%

Chemical Identifiers

106-95-6
120.977
BHELZAPQIKSEDF-UHFFFAOYSA-N
7841
C=CCBr
C3H5Br
MFCD00000244
allyl bromide, 3-bromopropene, 1-propene, 3-bromo, 3-bromopropylene, 3-bromo-1-propene, bromallylene, 2-propenyl bromide, 1-bromo-2-propene, propene, 3-bromo, allylbromide
3-bromoprop-1-ene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H301-H330-H340-H350-H314-H318
Highly flammable liquid and vapor.
Toxic if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H301+H331-H314-H335-H340-H350

DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: ALLYL BROMIDE

EINECSNumber : 203-446-6

RTECSNumber : UC7090000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only