missing translation for 'onlineSavingsMsg'
Learn More

Allyl Benzyl Ether 99.0+%, TCI America™

Catalog No. p-7117430
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A142725ML 25 mL
A1427100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A142725ML Supplier TCI America Supplier No. A142725ML

Chemical Identifiers

CAS 14593-43-2
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00078288
InChI Key HUGHWHMUUQNACD-UHFFFAOYSA-N
PubChem CID 84542
IUPAC Name prop-2-enoxymethylbenzene
SMILES C=CCOCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 205°C
Quantity 25 mL
Formula Weight 148.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Benzyl Ether