accessibility menu, dialog, popup

UserName

CAS RN 14593-43-2

H2CO

CAS RN 14593-43-2

IUPAC Name: [(prop-2-en-1-yloxy)methyl]benzene
Synonyms: [(prop-2-en-1-yloxy)methyl]benzene
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: HUGHWHMUUQNACD-UHFFFAOYSA-N
SMILES: C=CCOCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results