missing translation for 'onlineSavingsMsg'
Learn More

Allyl Benzoate 98.0+%, TCI America™

Catalog No. B204125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B204125G 25 g
B2041250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B204125G Supplier TCI America Supplier No. B204125G
Only null left

Chemical Identifiers

CAS 583-04-0
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00026104
InChI Key LYJHVEDILOKZCG-UHFFFAOYSA-N
Synonym Benzoic Acid Allyl Ester
PubChem CID 11406
IUPAC Name prop-2-enyl benzoate
SMILES C=CCOC(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 242°C
Quantity 25 g
Formula Weight 162.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Benzoate
EINECSNumber (3)-1357
TSCA No