accessibility menu, dialog, popup

UserName

H2COO

CAS RN 583-04-0

IUPAC Name: prop-2-en-1-yl benzoate
Synonyms: prop-2-en-1-yl benzoate
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: LYJHVEDILOKZCG-UHFFFAOYSA-N
SMILES: C=CCOC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results