missing translation for 'onlineSavingsMsg'
Learn More

Allyl 4-Hydroxybenzoate 98.0+%, TCI America™

Catalog No. A184625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A184625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A184625G Supplier TCI America Supplier No. A184625G
Only null left

Chemical Identifiers

CAS 18982-18-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD08276268
InChI Key NVNSVQJVBIWZNM-UHFFFAOYSA-N
Synonym Allylparaben, 4-Hydroxybenzoic Acid Allyl Ester
PubChem CID 11171300
IUPAC Name prop-2-en-1-yl 4-hydroxybenzoate
SMILES OC1=CC=C(C=C1)C(=O)OCC=C

Specifications

Melting Point 103°C
Color White-Yellow
Quantity 25 g
Formula Weight 178.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Allyl 4-Hydroxybenzoate
EINECSNumber (9)-1097
TSCA No