accessibility menu, dialog, popup

UserName

H2COOOH

CAS RN 18982-18-8

IUPAC Name: prop-2-en-1-yl 4-hydroxybenzoate
Synonyms: prop-2-en-1-yl 4-hydroxybenzoate
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: NVNSVQJVBIWZNM-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(=O)OCC=C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results