missing translation for 'onlineSavingsMsg'
Learn More

Allyl 1-Pyrrolidinecarbodithioate 95.0+%, TCI America™

Catalog No. P105625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P105625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P105625G Supplier TCI America Supplier No. P105625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 701-13-3
Molecular Formula C8H13NS2
Molecular Weight (g/mol) 187.319
MDL Number MFCD00135118
InChI Key GDQCWIUHBJJDNL-UHFFFAOYSA-N
Synonym Pyrrolidinodithiocarbamic Acid Allyl Ester, Allyl Pyrrolidinodithiocarbamate, 1-Pyrrolidinecarbodithioic Acid Allyl Ester
PubChem CID 575099
IUPAC Name prop-2-enyl pyrrolidine-1-carbodithioate
SMILES C=CCSC(=S)N1CCCC1

Specifications

Color Yellow
Quantity 25 g
Formula Weight 187.32
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl 1-Pyrrolidinecarbodithioate
TSCA No
Recommended Storage Refrigerator