accessibility menu, dialog, popup

UserName

H2CSSN

CAS RN 701-13-3

IUPAC Name: prop-2-en-1-yl pyrrolidine-1-carbodithioate
Synonyms: prop-2-en-1-yl pyrrolidine-1-carbodithioate
Molecular Weight (g/mol): 187.32
Molecular Formula: C8H13NS2
InChi Key: GDQCWIUHBJJDNL-UHFFFAOYSA-N
SMILES: C=CCSC(=S)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results