missing translation for 'onlineSavingsMsg'
Learn More

Troemner™ Alloy 8 Metric Precision Weight Sets, UltraClass

Excellent for high-precision calibrations and reference standards

$2954.71 - $5568.59

Specifications

Product Type Medium Metric Analytical Weight Set
Classification UltraClass
View More Specs

Products 4
Catalog Number Mfr. No. Mass Weight (Metric) Material Quantity Price Quantity  
Catalog Number Mfr. No. Mass Weight (Metric) Material Quantity Price Quantity  
02-214-052
View Documents
N/A 1 mg to 200 g 1 mg to 200 g Stainless Steel 22/Set
Each for $5,568.59
Only null left
 
02-214-3C
View Documents
Troemner™
30390073
1 mg to 100 g 1 mg to 100 g Stainless Steel, Aluminum 21/Set
Each for $4,370.88
Only null left
 
02-100-172
View Documents
Troemner™
30390072
1 to 50 g 1 to 50 g Stainless Steel 8/Set
Each for $2,954.71
Only null left
 
02-214-3B
View Documents
Troemner™
30390075
10 mg to 100 g 10 mg to 100 g Stainless Steel 17/Set
Each for $4,031.56
Only null left
 
Description

Description

Excellent for high-precision calibrations and reference standards

Manufactured from a specially developed austenitic stainless-steel—Alloy 8—available exclusively from Troemner. Weights 1g and larger are manufactured using Alloy 8, which possesses closely controlled density, extremely low magnetic susceptibility, excellent stability and resistance to corrosion, assuring a very stable mass over time. Other weights are stainless steel.

  • Each fractional weight is embossed with its value
  • Weights from 1mg to 30mg are aluminum and flat, with one side turned up to form a handle for non-slip gripping with forceps
  • Weights from 50 to 500mg are stainless steel and flat with one side turned up to form a handle
  • Gram and kilogram weights are manufactured using Alloy 8 and come in cylindrical shapes with knobs on top for easy handling
  • Weight sets come in a 5-2-2-1 series, with two weights of any included mg or g value beginning with 2quotation (2mg, 20mg, etc.) and one weight for all other inclusive values
  • Each weight set (100g to 1mg) is packaged in a polypropylene case
  • Weight sets 500g and above come packaged in a wood case
Specifications

Specifications

Medium Metric Analytical Weight Set
UltraClass
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'