Learn More
Troemner™ Alloy 8 Metric Precision Weight Sets, UltraClass
Excellent for high-precision calibrations and reference standards
Supplier: Troemner™ 30390075
Includes:
Forceps
Description
Excellent for high-precision calibrations and reference standards
Manufactured from a specially developed austenitic stainless-steel—Alloy 8—available exclusively from Troemner. Weights 1g and larger are manufactured using Alloy 8, which possesses closely controlled density, extremely low magnetic susceptibility, excellent stability and resistance to corrosion, assuring a very stable mass over time. Other weights are stainless steel.
- Each fractional weight is embossed with its value
- Weights from 1mg to 30mg are aluminum and flat, with one side turned up to form a handle for non-slip gripping with forceps
- Weights from 50 to 500mg are stainless steel and flat with one side turned up to form a handle
- Gram and kilogram weights are manufactured using Alloy 8 and come in cylindrical shapes with knobs on top for easy handling
- Weight sets come in a 5-2-2-1 series, with two weights of any included mg or g value beginning with 2quotation (2mg, 20mg, etc.) and one weight for all other inclusive values
- Each weight set (100g to 1mg) is packaged in a polypropylene case
- Weight sets 500g and above come packaged in a wood case
Certifications
A NIST/NVLAP™ weight calibration certificate or traceable certificate may be purchased to meet all of your ISO™, GLP™, GMP™, FDA, or nuclear requirements.
Specifications
| Medium Metric Analytical Weight Set | |
| Stainless Steel | |
| 10 mg to 100 g | |
| 17/Set |
| UltraClass | |
| 10 mg to 100 g | |
| Cylindrical weight: 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, Polycarbonate case | |
| Multiple |