missing translation for 'onlineSavingsMsg'
Learn More
Learn More
all-trans-Retinal, Spectrum™ Chemical
R3041, 116-31-4, C20H28O
$418.73 - $395864.61
Chemical Identifiers
| CAS | 116-31-4 |
|---|---|
| Molecular Formula | C20H28O |
| Molecular Weight (g/mol) | 284.44 |
| InChI Key | NCYCYZXNIZJOKI-OVSJKPMPSA-N |
| IUPAC Name | (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenal |
| SMILES | C\C(\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C)=C/C=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18600415
|
Spectrum Chemical Mfg Cor
R3041100MG |
100 mg | Amber Glass Bottle |
Each for $418.73
|
|
||||
|
18600416
|
Spectrum Chemical Mfg Cor
R3041250MG |
250 mg | Amber Glass Bottle |
Each for $634.00
|
|
||||
|
18600417
|
Spectrum Chemical Mfg Cor
R30411GM |
1 g | Amber Glass Bottle |
Each for $1,236.42
|
|
||||
|
18600418
|
Spectrum Chemical Mfg Cor
R30415GM |
5 g | Amber Glass Bottle |
Each for $4,939.24
|
|
||||
|
18600419
|
Spectrum Chemical Mfg Cor
R3041100GMBL |
100 g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18600420
|
Spectrum Chemical Mfg Cor
R3041160GMBL |
160 g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18600421
|
Spectrum Chemical Mfg Cor
R3041390GMBL |
390 g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical all-trans-Retinal is also known as retinaldehyde and considered a form of vitamin A. It is the chemical basis of animal vision. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
| 116-31-4 | |
| 284.44 | |
| (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenal |
| C20H28O | |
| NCYCYZXNIZJOKI-OVSJKPMPSA-N | |
| C\C(\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C)=C/C=O |
Specifications
| 116-31-4 | |
| Amber Glass Bottle | |
| 100 mg | |
| C\C(\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C)=C/C=O | |
| 284.44 |
| 100% | |
| C20H28O | |
| NCYCYZXNIZJOKI-OVSJKPMPSA-N | |
| (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenal | |
| Reagent |