missing translation for 'onlineSavingsMsg'
Learn More

all-trans-Retinal, Spectrum™ Chemical

Catalog No. 18600415
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
100 g
160 g
390 g
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600415 100 mg Amber Glass Bottle
18600416 250 mg Amber Glass Bottle
18600417 1 g Amber Glass Bottle
18600418 5 g Amber Glass Bottle
18600419 100 g Amber Glass Bottle
18600420 160 g Amber Glass Bottle
18600421 390 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18600415 Supplier Spectrum Chemical Mfg Cor Supplier No. R3041100MG

R3041, 116-31-4, C20H28O

Spectrum™ Chemical all-trans-Retinal is also known as retinaldehyde and considered a form of vitamin A. It is the chemical basis of animal vision. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 116-31-4
Molecular Formula C20H28O
Molecular Weight (g/mol) 284.44
InChI Key NCYCYZXNIZJOKI-OVSJKPMPSA-N
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenal
SMILES C\C(\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C)=C/C=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 mg
Grade Reagent