missing translation for 'onlineSavingsMsg'
Learn More
Learn More
all-trans-Retinal, Spectrum™ Chemical
R3041, 116-31-4, C20H28O
Supplier: Spectrum Chemical Mfg Cor R30411GM
Description
Spectrum™ Chemical all-trans-Retinal is also known as retinaldehyde and considered a form of vitamin A. It is the chemical basis of animal vision. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Specifications
| 116-31-4 | |
| Amber Glass Bottle | |
| 1 g | |
| C\C(\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C)=C/C=O | |
| 284.44 |
| 100% | |
| C20H28O | |
| NCYCYZXNIZJOKI-OVSJKPMPSA-N | |
| (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenal | |
| Reagent |
Chemical Identifiers
| 116-31-4 | |
| 284.44 | |
| (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenal |
| C20H28O | |
| NCYCYZXNIZJOKI-OVSJKPMPSA-N | |
| C\C(\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C)=C/C=O |