missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Alkaline Phosphatase Buffer-1 (5X)
For cellular and molecular biology applications
$160.68
Specifications
| Chemical Name or Material | Alkaline Phosphatase Buffer-1 (5X) |
|---|---|
| Physical Form | Liquid |
| Formulation | 200mM Tris, 200mM NaCl, and 40mM magnesium chloride at pH 9.5 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ62907AK
|
Thermo Scientific™
J62907AK |
250 mL |
Each for $160.68
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Formulation: 200mM Tris, 200mM NaCl, and 40mM magnesium chloride at pH 9.5
Specifications
| Alkaline Phosphatase Buffer-1 (5X) | |
| Liquid | |
| 200mM Tris, 200mM NaCl, and 40mM magnesium chloride at pH 9.5 |
Safety and Handling
Recommended Storage : Keep cold
For Research Use Only