missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Alkaline Phosphatase Buffer-1 (5X)

Catalog No. AAJ62907AK
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62907AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62907AK Supplier Thermo Scientific™ Supplier No. J62907AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For cellular and molecular biology applications

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Formulation: 200mM Tris, 200mM NaCl, and 40mM magnesium chloride at pH 9.5

Specifications

Chemical Name or Material Alkaline Phosphatase Buffer-1 (5X)
Physical Form Liquid
Quantity 250 mL
Formulation 200mM Tris, 200mM NaCl, and 40mM magnesium chloride at pH 9.5

For Research Use Only