missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alizarin Red S Indicator, 1% (w/v), Ricca Chemical
$61.97 - $191.87
Chemical Identifiers
| CAS | 130-22-3 |
|---|---|
| Molecular Formula | C14H7NaO7S |
| Molecular Weight (g/mol) | Mixture |
| InChI Key | HFVAFDPGUJEFBQ-UHFFFAOYSA-M |
| PubChem CID | 3955344 |
| ChEBI | CHEBI:87358 |
| IUPAC Name | sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate |
| SMILES | C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
R5004
|
Ricca Chemical Company
RI5004 |
120 mL | Natural Poly Bottle |
Each for $61.97
|
|
||||
|
500-16
|
Ricca Chemical Company
RI50016 |
500 mL | Natural Poly Bottle |
Each for $110.83
|
|
||||
|
50032
|
Ricca Chemical Company
50032 |
1 L | Natural Poly Bottle |
Each for $191.87
|
|
||||
Chemical Identifiers
| 130-22-3 | |
| Mixture | |
| 3955344 | |
| sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate |
| C14H7NaO7S | |
| HFVAFDPGUJEFBQ-UHFFFAOYSA-M | |
| CHEBI:87358 | |
| C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+] |
Specifications
| 130-22-3 , 7732-18-5 | |
| 100.0,1.01 | |
| C14H7NaO7S | |
| C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+] | |
| Mixture | |
| CHEBI:87358 | |
| Indicator | |
| Aqueous Solution | |
| 4.5 to 5 | |
| Liquid |
| 97.0,0.97 | |
| 1% (w/v) | |
| HFVAFDPGUJEFBQ-UHFFFAOYSA-M | |
| sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate | |
| 3955344 | |
| 1% W/V | |
| Natural Poly Bottle | |
| Dark Red | |
| 120 mL | |
| Alizarin Red S Indicator |