missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alizarin Cyanin Green F, TCI America™
Supplier: TCI America A061025G
Specifications
| Alizarin Cyanin Green F | |
| 238°C | |
| C28H20N2Na2O8S2 | |
| 25 g | |
| FPAYXBWMYIMERV-UHFFFAOYSA-L | |
| disodium;5-methyl-2-[[4-(4-methyl-2-sulfonatoanilino)-9,10-dioxoanthracen-1-yl]amino]benzenesulfonate | |
| 20431 | |
| Crystalline Powder |
| 4403-90-1 | |
| Black-Green | |
| MFCD00001193 | |
| Acid Green 25 | |
| CC1=CC(=C(C=C1)NC2=C3C(=C(C=C2)NC4=C(C=C(C=C4)C)S(=O)(=O)[O-])C(=O)C5=CC=CC=C5C3=O)S(=O)(=O)[O-].[Na+].[Na+] | |
| 622.574 | |
| 622.57 |
Chemical Identifiers
| 4403-90-1 | |
| 622.574 | |
| FPAYXBWMYIMERV-UHFFFAOYSA-L | |
| 20431 | |
| CC1=CC(=C(C=C1)NC2=C3C(=C(C=C2)NC4=C(C=C(C=C4)C)S(=O)(=O)[O-])C(=O)C5=CC=CC=C5C3=O)S(=O)(=O)[O-].[Na+].[Na+] |
| C28H20N2Na2O8S2 | |
| MFCD00001193 | |
| Acid Green 25 | |
| disodium;5-methyl-2-[[4-(4-methyl-2-sulfonatoanilino)-9,10-dioxoanthracen-1-yl]amino]benzenesulfonate |
Safety and Handling
EINECSNumber : (5)-1741
RTECSNumber : DB5044000
TSCA : Yes