missing translation for 'onlineSavingsMsg'
Learn More

Aliphatic Hydrocarbons Kit, MilliporeSigma™ Supelco™

Catalog No. 111016536
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016536 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016536 Supplier Merck Emd Millipore Supplier No. 44575U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Petroleum

Specifications

CAS 110-54-3
Flash Point -17°C
Type Analytical Standard
Quantity 1 Ea.
Grade Analytical Standard