Learn More
Thermo Scientific™ aLICator LIC Cloning and Expression Kit 1 (untagged)
Get fast and efficient ligation-independent cloning and tight regulation of gene expression in E. coli. with this cloning and expression system.
Supplier: Thermo Scientific™ K1241
Description
aLICator LIC Cloning and Expression System is designed for fast and efficient ligation independent cloning and tight regulation of gene expression in E. coli. The pLATE bacterial expression vectors are designed for high levels of target protein expression in concert with minimal background (uninduced) expression, which permits expression of proteins that are toxic to E. coli cells. To streamline and facilitate the process of insert cloning into the expression vector, the aLICator system uses directional LIC cloning technology, a rapid procedure that provides high-cloning efficiencies.
The tightly regulated expression and fast, efficient directional cloning makes the aLICator LIC Cloning and Expression System the best choice for routine and toxic gene cloning and expression in E. coli.
Highlights
- High efficiency LIC cloning
- Tight control of gene expression
- High yield expression
- Versatile -tagged or untaged protein expression with tag removal option
Specifications
| Ampicillin (AmpR) | |
| Kit | |
| PCR Cloning | |
| Cloning and Expression Kit | |
| Untagged | |
| pLATE |
| LIC Cloning | |
| PCR Products | |
| aLICator | |
| T7 | |
| 20 Reactions |
For Research Use Only. Not for use in diagnostic procedures.